Gene Transcripts

Gene Symbol
UST
Entrez Gene ID
10090
Gene Name
uronyl 2-sulfotransferase
Gene Aliases
2OST
Chromosome Location
Chr. 6: 148745891 - 149076990 on Build GRCh38
UniGene ID
Hs.657370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005715.2 NP_005706.1 3 708
XM_011535378.2 XP_011533680.1 3 1399
XM_017010152.1 XP_016865641.1 3 1952
GenBank mRNA AB020316.1 - 3 515
AK292922.1 - 3 456
AK315320.1 - 3 416
BC093668.1 - 3 460 7939513
BC093694.1 - 3 460 7939539

View Full Product Details