Gene Transcripts

Gene Symbol
LRRC45
Entrez Gene ID
201255
Gene Name
leucine rich repeat containing 45
Gene Aliases
Chromosome Location
Chr. 17: 82023252 - 82031151 on Build GRCh38
UniGene ID
Hs.143774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144999.3 NP_659436.1 3, 4 689
GenBank mRNA BC014109.2 - 3, 4 587
BI837975.1 - 3, 4 658

View Full Product Details