Gene Transcripts

Gene Symbol
EFCAB8
Entrez Gene ID
388795
Gene Name
EF-hand calcium binding domain 8
Gene Aliases
Chromosome Location
Chr. 20: 32860995 - 32961845 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011529121.1 XP_011527423.1 23 2960

View Full Product Details