Gene Transcripts
- Gene Symbol
- ZDHHC11
- Entrez Gene ID
- 79844
- Gene Name
- zinc finger DHHC-type containing 11
- Gene Aliases
- ZNF399
- Chromosome Location
- Chr. 5: 795605 - 863634 on Build GRCh38
- UniGene ID
- Hs.654874
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096220.1 |
- |
|
2464 |
|
- Gene Symbol
- ZDHHC11B
- Entrez Gene ID
- 653082
- Gene Name
- zinc finger DHHC-type containing 11B
- Gene Aliases
- DHHC-11B
- Chromosome Location
- Chr. 5: 710355 - 784749 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details