Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177576.1 | NP_001171047.1 | 10 | 1784 | |
NM_026646.3 | NP_080922.1 | 10 | 2026 | ||
GenBank mRNA | AK010193.1 | - | 10 | 1772 | |
AK046874.1 | - | 10 | 1773 | ||
AK048389.1 | - | 10 | 1755 | ||
AK049313.1 | - | 10 | 1755 | ||
AK155276.1 | - | 10 | 1778 | ||
AK158985.1 | - | 1 | 4832 | ||
AK162400.1 | - | 10 | 1738 | ||
BC037949.1 | - | 10 | 1783 | 5367108 | |
BC050887.1 | - | 9 | 2502 |