Gene Transcripts

Gene Symbol
Pxt1
Entrez Gene ID
69307
Gene Name
peroxisomal, testis specific 1
Gene Aliases
1700001G18Rik, Stepp
Chromosome Location
Chr. 17: 28933986 - 28942262 on Build GRCm38
UniGene ID
Mm.107605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153390.1 NP_700439.1 3 961
GenBank mRNA AF486826.1 - 3 961
AK005593.1 - 3 962
BC100437.1 - 3 960 30915792

View Full Product Details