Gene Transcripts

Gene Symbol
Clic3
Entrez Gene ID
69454
Gene Name
chloride intracellular channel 3
Gene Aliases
2300003G24Rik
Chromosome Location
Chr. 2: 25456843 - 25458776 on Build GRCm38
UniGene ID
Mm.44194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027085.3 NP_081361.1 5 565
GenBank mRNA AK009020.1 - 5 567
BC060967.1 - 5 570 30297308

View Full Product Details