Gene Transcripts

Gene Symbol
LZTS1
Entrez Gene ID
11178
Gene Name
leucine zipper tumor suppressor 1
Gene Aliases
F37, FEZ1
Chromosome Location
Chr. 8: 20246165 - 20303981 on Build GRCh38
UniGene ID
Hs.521432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021020.3 NP_066300.1 3 1733
XM_005273394.3 XP_005273451.1 4 1998
XM_011544383.2 XP_011542685.1 4 3936
XM_011544384.2 XP_011542686.1 4 1886
XM_011544385.2 XP_011542687.1 4 1860
XM_011544386.2 XP_011542688.1 4 2085
GenBank mRNA AF123655.1 - 4 1445
AF123657.1 - 4 1523
AF123658.1 - 4 1553
AF123659.1 - 3 1733
BC075006.2 - 3 1671 30915401
BC075007.2 - 3 1671 30915614

View Full Product Details