Gene Transcripts

Gene Symbol
SLC2A6
Entrez Gene ID
11182
Gene Name
solute carrier family 2 member 6
Gene Aliases
GLUT6, GLUT9, HSA011372
Chromosome Location
Chr. 9: 133471094 - 133479154 on Build GRCh38
UniGene ID
Hs.244378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145099.1 NP_001138571.1 2 246
NM_017585.3 NP_060055.2 2 246
XM_011518189.2 XP_011516491.2 2 219
XM_017014236.1 XP_016869725.1 2 228
XM_017014237.1 XP_016869726.1 2 219
XM_017014238.1 XP_016869727.1 2 219
GenBank mRNA AB209108.1 - 1 511
AJ011372.2 - 2 188
AK074836.1 - 2 196
AK074927.1 - 2 229
AK096630.1 - 1 749
AK222919.1 - 2 191
AK225106.1 - 2 188
AK294974.1 - 2 166
BC013740.1 - 2 180 4449594
DC305025.1 - 2 246
Y17803.1 - 2 197

View Full Product Details