Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204514.1 | NP_001191443.1 | 19 | 2540 | |
NM_001204516.1 | NP_001191445.1 | 17 | 2244 | ||
NM_001204517.1 | NP_001191446.1 | 18 | 3485 | ||
NM_014970.3 | NP_055785.2 | 18 | 2376 | ||
XM_005244970.1 | XP_005245027.1 | 18 | 2379 | ||
XM_011509307.2 | XP_011507609.2 | 18 | 2379 | ||
GenBank mRNA | AK026471.1 | - | 18 | 2155 | |
AK122897.1 | - | 18 | 3487 | ||
AK297368.1 | - | 17 | 2171 | ||
AK301912.1 | - | 11 | 1343 | ||
AK303052.1 | - | 19 | 2497 | ||
AK315586.1 | - | 18 | 2290 | ||
BC028679.2 | - | 18 | 2333 | 4829354 | |
BC048012.1 | - | 17 | 2288 | ||
U59919.1 | - | 18 | 2293 |