Gene Transcripts
- Gene Symbol
- MALRD1
- Entrez Gene ID
- 340895
- Gene Name
- MAM and LDL receptor class A domain containing 1
- Gene Aliases
- C10orf112, DIET1, bA265G8.2
- Chromosome Location
- Chr. 10: 19046931 - 19734478 on Build GRCh38
- UniGene ID
- Hs.646734
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142308.2 |
NP_001135780.2
|
38, 39 |
6475 |
|
XM_011519453.2 |
XP_011517755.1
|
39, 40 |
7013 |
|
XM_011519454.1 |
XP_011517756.1
|
39, 40 |
6339 |
|
XM_011519455.2 |
XP_011517757.1
|
38, 39 |
6840 |
|
XM_017016182.1 |
XP_016871671.1
|
38, 39 |
6285 |
|
XM_017016184.1 |
XP_016871673.1
|
27, 28 |
4702 |
|
XM_017016185.1 |
XP_016871674.1
|
27, 28 |
4699 |
|
XM_017016186.1 |
XP_016871675.1
|
20, 21 |
3256 |
|
GenBank mRNA |
AK297683.1 |
- |
10, 11 |
1358 |
|
KC843478.1 |
- |
38, 39 |
6475 |
|
View Full Product Details