Gene Transcripts

Gene Symbol
DYTN
Entrez Gene ID
391475
Gene Name
dystrotelin
Gene Aliases
Chromosome Location
Chr. 2: 206651621 - 206718396 on Build GRCh38
UniGene ID
Hs.640667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001093730.1 NP_001087199.1 11 1730
GenBank mRNA DQ516347.1 - 11 1730

View Full Product Details