Gene Transcripts

Gene Symbol
LSM14B
Entrez Gene ID
149986
Gene Name
LSM family member 14B
Gene Aliases
C20orf40, FAM61B, FT005, LSM13, RAP55B, bA11M20.3
Chromosome Location
Chr. 20: 62122454 - 62135378 on Build GRCh38
UniGene ID
Hs.105379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144703.2 NP_653304.2 2 398
XM_005260302.3 XP_005260359.1 2 393
XM_011528605.2 XP_011526907.1 2 399
XM_011528606.2 XP_011526908.1 2 395
XM_011528607.2 XP_011526909.1 2 403
XM_011528608.2 XP_011526910.1 2 396
XM_011528609.2 XP_011526911.1 2 394
XM_011528611.2 XP_011526913.1 2 394
XM_011528612.2 XP_011526914.1 2 147
XM_011528613.2 XP_011526915.1 2 115
XM_011528614.2 XP_011526916.1 2 328
XM_011528615.2 XP_011526917.1 1 65
XM_017027688.1 XP_016883177.1 2 395
XM_017027689.1 XP_016883178.1 2 392
XM_017027690.1 XP_016883179.1 1 65
GenBank mRNA AK058202.1 - 2 152
BC035702.1 - 2 385
BC057387.1 - 2 398 5724236
BM547301.1 - 2 398
BX375905.2 - 2 382

View Full Product Details