Gene Transcripts

Gene Symbol
C1orf68
Entrez Gene ID
100129271
Gene Name
chromosome 1 open reading frame 68
Gene Aliases
LEP7, XP32
Chromosome Location
Chr. 1: 152719522 - 152720429 on Build GRCh38
UniGene ID
Hs.601077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024679.2 NP_001019850.1 1 293
GenBank mRNA CU446285.1 - 1 133
CU457096.1 - 1 256

View Full Product Details