Gene Transcripts

Gene Symbol
Gm11565
Entrez Gene ID
670550
Gene Name
predicted gene 11565
Gene Aliases
OTTMUSG00000002203
Chromosome Location
Chr. 11: 99914751 - 99915671 on Build GRCm38
UniGene ID
Mm.379775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126323.1 NP_001119795.1 1 764

View Full Product Details