Gene Transcripts
- Gene Symbol
- Tbc1d4
- Entrez Gene ID
- 210789
- Gene Name
- TBC1 domain family, member 4
- Gene Aliases
- 5930406J04Rik, A930035N22, AV295684, As160
- Chromosome Location
- Chr. 14: 101442360 - 101609228 on Build GRCm38
- UniGene ID
- Mm.320639
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081278.2 |
NP_001074747.2
|
7, 8 |
2369 |
|
GenBank mRNA |
AK031120.1 |
- |
7, 8 |
2213 |
|
AK039337.1 |
- |
7, 8 |
2370 |
|
AK044719.1 |
- |
7, 8 |
2011 |
|
AK220347.1 |
- |
6, 7 |
776 |
|
BC042747.1 |
- |
3, 4 |
371 |
|
BC064433.1 |
- |
7, 8 |
1993 |
|
BC080762.1 |
- |
7, 8 |
1987 |
|
BC117869.1 |
- |
7, 8 |
1842 |
40095526 |
BC117870.1 |
- |
7, 8 |
1845 |
40095531 |
View Full Product Details