Gene Transcripts

Gene Symbol
Cox17
Entrez Gene ID
89786
Gene Name
COX17 cytochrome c oxidase copper chaperone
Gene Aliases
Chromosome Location
Chr. 11: 64962663 - 64968437 on Build Rnor_6.0
UniGene ID
Rn.19207
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053540.2 NP_445992.1 2 233
GenBank mRNA AB032178.1 - 2 177

View Full Product Details