Gene Transcripts

Gene Symbol
Tmem14c
Entrez Gene ID
171432
Gene Name
transmembrane protein 14C
Gene Aliases
Cdtw1
Chromosome Location
Chr. 17: 21571510 - 21577652 on Build Rnor_6.0
UniGene ID
Rn.114642
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135169.2 NP_001128641.1 5 380
NM_134395.2 NP_599222.2 4 593
XM_006253777.2 XP_006253839.1 5 583
GenBank mRNA AF083396.1 - 4 601
BC158854.1 - 4 274 7936967
CB744788.1 - 5 434
FQ211234.1 - 5 370
FQ217516.1 - 5 370
FQ218209.1 - 5 376
FQ223586.1 - 5 360
FQ223742.1 - 5 370
FQ224495.1 - 5 376
FQ225707.1 - 5 350
FQ230083.1 - 5 398
FQ230323.1 - 5 379
FQ231364.1 - 5 360
FQ233821.1 - 5 368

View Full Product Details