Gene Transcripts

Gene Symbol
INTS1
Entrez Gene ID
26173
Gene Name
integrator complex subunit 1
Gene Aliases
INT1, NET28
Chromosome Location
Chr. 7: 1470277 - 1504382 on Build GRCh38
UniGene ID
Hs.532188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080453.2 NP_001073922.2 10 1486
XM_011515260.1 XP_011513562.1 10 1486
XM_011515262.2 XP_011513564.1 10 1486
XM_017011959.1 XP_016867448.1 10 1486
XM_017011960.1 XP_016867449.1 10 1486
GenBank mRNA BF346658.1 - 1 21
BM547069.1 - 4 496
BM553299.1 - 7 915

View Full Product Details