Gene Transcripts

Gene Symbol
ATP2C1
Entrez Gene ID
27032
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location
Chr. 3: 130850500 - 131016712 on Build GRCh38
UniGene ID
Hs.584884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001485.2 NP_001001485.1 25 2798
NM_001001486.1 NP_001001486.1 24 2475
NM_001001487.1 NP_001001487.1 24 2475
NM_001199179.1 NP_001186108.1 25 2704
NM_001199180.1 NP_001186109.1 24 2693
NM_001199181.1 NP_001186110.1 24 2693
NM_001199182.1 NP_001186111.1 23 2576
NM_001199183.1 NP_001186112.1 24 2566
NM_001199184.1 NP_001186113.1 24 2566
NM_001199185.1 NP_001186114.1 24 2617
NM_014382.3 NP_055197.2 24 2617
XM_005247354.2 XP_005247411.1 24 2718
XM_005247355.2 XP_005247412.1 25 2845
XM_005247356.2 XP_005247413.1 25 2598
XM_005247358.2 XP_005247415.1 24 2422
XM_011512686.2 XP_011510988.1 15 1527
XM_017006164.1 XP_016861653.1 25 2506
GenBank mRNA AB037768.1 - 25 2705
AB209265.1 - 13 2465
AF181120.1 - 24 2530
AF181121.1 - 24 2530
AF189723.2 - 24 2449
AF225981.1 - 24 2449
AJ010953.1 - 10 1027
AK001684.1 - 12 1225
AK074692.1 - 22 2156
AK296470.1 - 24 2620
AK299945.1 - 23 2332
AK304175.1 - 24 2422
AK304374.1 - 24 2449
AK314342.1 - 24 2479
AY268374.1 - 24 2424
AY268375.1 - 24 2424
BC028139.1 - 24 2487 5215224

View Full Product Details