Gene Transcripts

Gene Symbol
SND1
Entrez Gene ID
27044
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
TDRD11, p100
Chromosome Location
Chr. 7: 127652119 - 128092607 on Build GRCh38
UniGene ID
Hs.122523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014390.2 NP_055205.2 3 513
XM_017011987.1 XP_016867476.1 3 542
GenBank mRNA AB209510.1 - 3 451
AK096583.1 - 4 438
AK304181.1 - 3 468
AK312313.1 - 3 467
BC017180.2 - 3 469 3345037
BT009785.1 - 3 287
U22055.1 - 3 479

View Full Product Details