Gene Transcripts

Gene Symbol
SAC3D1
Entrez Gene ID
29901
Gene Name
SAC3 domain containing 1
Gene Aliases
HSU79266, SHD1
Chromosome Location
Chr. 11: 65040904 - 65044828 on Build GRCh38
UniGene ID
Hs.23642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013299.3 NP_037431.3 2 1515
GenBank mRNA U79266.1 - 2 1510

View Full Product Details