Gene Transcripts

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
Chromosome Location
Chr. 19: 47608196 - 47703277 on Build GRCh38
UniGene ID
Hs.97244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015711.3 NP_056526.3 8 3073
XM_005258833.4 XP_005258890.1 8 3116
XM_011526882.2 XP_011525184.1 4 4083
XM_011526883.2 XP_011525185.1 7 2939
GenBank mRNA AF182077.1 - 3 3089

View Full Product Details