Gene Transcripts
- Gene Symbol
- CDK11B
- Entrez Gene ID
- 984
- Gene Name
- cyclin dependent kinase 11B
- Gene Aliases
- CDC2L1, CDK11, CDK11-p110, CDK11-p46, CDK11-p58, CLK-1, PITSLREA, PK58, p58, p58CDC2L1, p58CLK-1
- Chromosome Location
- Chr. 1: 1635226 - 1659839 on Build GRCh38
- UniGene ID
- Hs.651228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291345.1 |
NP_001278274.1
|
9, 10 |
1175 |
|
NM_001787.2 |
NP_001778.2
|
9, 10 |
1244 |
|
NM_033486.2 |
NP_277021.2
|
9, 10 |
1205 |
|
NM_033487.2 |
NP_277022.1
|
7, 8 |
945 |
|
NM_033489.2 |
NP_277024.2
|
10, 11 |
1252 |
|
NM_033490.2 |
NP_277025.1
|
8, 9 |
1068 |
|
XM_006711065.3 |
XP_006711128.1
|
9, 10 |
1956 |
|
XM_011542490.2 |
XP_011540792.1
|
9, 10 |
2013 |
|
XM_011542491.2 |
XP_011540793.1
|
9, 10 |
1469 |
|
XM_011542492.2 |
XP_011540794.1
|
9, 10 |
1320 |
|
XM_011542493.2 |
XP_011540795.1
|
9, 10 |
1983 |
|
XM_011542494.2 |
XP_011540796.1
|
10, 11 |
2060 |
|
XM_011542495.2 |
XP_011540797.1
|
10, 11 |
2030 |
|
XM_011542496.2 |
XP_011540798.1
|
4, 5 |
500 |
|
XM_017002925.1 |
XP_016858414.1
|
9, 10 |
1974 |
|
XM_017002926.1 |
XP_016858415.1
|
9, 10 |
1944 |
|
XM_017002927.1 |
XP_016858416.1
|
9, 10 |
1936 |
|
XM_017002928.1 |
XP_016858417.1
|
10, 11 |
2033 |
|
GenBank mRNA |
AF067512.1 |
- |
9, 10 |
1119 |
|
AF067513.1 |
- |
7, 8 |
859 |
|
AF067514.1 |
- |
10, 11 |
1133 |
|
AF067515.1 |
- |
9, 10 |
1158 |
|
AF067516.1 |
- |
10, 11 |
1166 |
|
AF067517.1 |
- |
8, 9 |
982 |
|
AF174497.1 |
- |
3, 4 |
358 |
|
AK000081.1 |
- |
8, 9 |
1003 |
|
AK304679.1 |
- |
9, 10 |
1083 |
|
BC139777.1 |
- |
3, 4 |
288 |
|
BC140714.1 |
- |
9, 10 |
1078 |
9021582 |
BC171773.1 |
- |
9, 10 |
1078 |
9054427 |
U04816.1 |
- |
9, 10 |
1104 |
|
U04817.1 |
- |
9, 10 |
1077 |
|
U04818.1 |
- |
8, 9 |
973 |
|
U04824.1 |
- |
9, 10 |
1110 |
|
- Gene Symbol
- CDK11A
- Entrez Gene ID
- 728642
- Gene Name
- cyclin dependent kinase 11A
- Gene Aliases
- CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
- Chromosome Location
- Chr. 1: 1702383 - 1724565 on Build GRCh38
- UniGene ID
- Hs.651228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001313896.1 |
NP_001300825.1
|
9, 10 |
1332 |
|
NM_001313982.1 |
NP_001300911.1
|
9, 10 |
1320 |
|
NM_024011.3 |
NP_076916.2
|
9, 10 |
1323 |
|
NM_033529.3 |
NP_277071.2
|
9, 10 |
1293 |
|
NR_132739.1 |
|
9, 10 |
1323 |
|
NR_132740.1 |
|
9, 10 |
1362 |
|
GenBank mRNA |
AF067518.1 |
- |
9, 10 |
1072 |
|
AF067519.1 |
- |
9, 10 |
1072 |
|
AF067520.1 |
- |
9, 10 |
1069 |
|
AF067521.1 |
- |
9, 10 |
1042 |
|
AF067522.1 |
- |
9, 10 |
1081 |
|
AF067523.1 |
- |
9, 10 |
1111 |
|
AK301495.1 |
- |
8, 9 |
918 |
|
AK303446.1 |
- |
9, 10 |
1081 |
|
AK303451.1 |
- |
9, 10 |
1064 |
|
AL043340.1 |
- |
2, 3 |
250 |
|
BC014464.1 |
- |
1, 2 |
54 |
|
BC094827.1 |
- |
1, 2 |
87 |
|
BC110905.1 |
- |
4, 5 |
400 |
6151813 |
U07704.1 |
- |
9, 10 |
1104 |
|
U07705.1 |
- |
9, 10 |
1098 |
|
View Full Product Details