Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001135998.2 | NP_001129470.1 | 1 | 721 | |
NM_019056.6 | NP_061929.2 | 1 | 721 | ||
GenBank mRNA | AF044213.1 | - | 1 | 208 | |
AF134476.1 | - | 1 | 255 | ||
AF251063.1 | - | 1 | 554 | ||
AK000501.1 | - | 1 | 203 | ||
AY359056.1 | - | 1 | 721 | ||
BC010665.1 | - | 1 | 529 | 3870660 | |
BC107805.1 | - | 1 | 204 | 6496406 | |
CR457239.1 | - | 1 | 190 |