Gene Transcripts

Gene Symbol
FAM63B
Entrez Gene ID
54629
Gene Name
family with sequence similarity 63 member B
Gene Aliases
Chromosome Location
Chr. 15: 58771192 - 58861900 on Build GRCh38
UniGene ID
Hs.591122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040450.2 NP_001035540.1 7 1652
NM_001040453.2 NP_001035543.1 7 1482
XM_011521687.1 XP_011519989.1 8 1704
XM_011521688.1 XP_011519990.1 8 1704
XM_011521689.1 XP_011519991.1 8 1602
XM_011521690.1 XP_011519992.1 7 1548
XM_017022332.1 XP_016877821.1 6 1458
XM_017022333.1 XP_016877822.1 5 1404
XM_017022334.1 XP_016877823.1 5 1404
GenBank mRNA AB032990.1 - 7 759
AK075319.1 - 7 1530
AK297291.1 - 7 1530
AL833610.1 - 6 1510
BC140811.1 - 7 1482 9021679
BC144616.1 - 7 1482 9053147

View Full Product Details