Gene Transcripts

Gene Symbol
HAUS6
Entrez Gene ID
54801
Gene Name
HAUS augmin like complex subunit 6
Gene Aliases
Dgt6, FAM29A
Chromosome Location
Chr. 9: 19053137 - 19102942 on Build GRCh38
UniGene ID
Hs.533468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270890.1 NP_001257819.1 6 889
NM_017645.4 NP_060115.3 6 889
XM_017014830.1 XP_016870319.1 2 40
GenBank mRNA AF533709.1 - 6 1071
AF537091.1 - 6 851
AK022964.1 - 3 205
AK056458.1 - 6 851
AL832495.1 - 6 849
BC026178.1 - 6 852
BC065935.1 - 4 389
BC111041.1 - 2 165 5529130
BC113981.1 - 6 686 40082296
BC114492.1 - 6 686 40082291

View Full Product Details