Gene Transcripts
- Gene Symbol
- HAUS6
- Entrez Gene ID
- 54801
- Gene Name
- HAUS augmin like complex subunit 6
- Gene Aliases
- Dgt6, FAM29A
- Chromosome Location
- Chr. 9: 19053137 - 19102942 on Build GRCh38
- UniGene ID
- Hs.533468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB046794.1 |
- |
4 |
362 |
|
AF537091.1 |
- |
11 |
1468 |
|
AL832495.1 |
- |
11 |
1466 |
|
View Full Product Details