Gene Transcripts

Gene Symbol
LAMTOR1
Entrez Gene ID
55004
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Gene Aliases
C11orf59, PDRO, Ragulator1, p18, p27RF-Rho
Chromosome Location
Chr. 11: 72097292 - 72103387 on Build GRCh38
UniGene ID
Hs.731528
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017907.2 NP_060377.1 5 605
GenBank mRNA AF289583.1 - 5 496
AK000632.1 - 5 494
AK300681.1 - 5 460
BC001706.1 - 5 467 3544000
BX381569.2 - 5 605
CR457247.1 - 5 442

View Full Product Details