Gene Transcripts

Gene Symbol
PIDD1
Entrez Gene ID
55367
Gene Name
p53-induced death domain protein 1
Gene Aliases
LRDD, PIDD
Chromosome Location
Chr. 11: 799179 - 809504 on Build GRCh38
UniGene ID
Hs.592290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145886.3 NP_665893.2 3 748
NM_145887.3 NP_665894.2 3 748
XM_005253005.4 XP_005253062.1 4 1686
XM_005253006.4 XP_005253063.1 4 1249
XM_005253007.3 XP_005253064.1 3 766
XM_005253008.4 XP_005253065.1 4 1746
XM_011520209.2 XP_011518511.1 3 984
XM_011520210.2 XP_011518512.1 4 1686
XM_011520211.2 XP_011518513.1 4 1686
XM_011520212.1 XP_011518514.1 3 826
GenBank mRNA AB208832.1 - 2 1270
AB208949.1 - 2 1234
AB209529.1 - 2 1376
AF229178.1 - 2 950
AF274972.1 - 3 744
BC014904.1 - 2 686 3908102
BX349115.2 - 1 101

View Full Product Details