Gene Transcripts

Gene Symbol
ZDHHC7
Entrez Gene ID
55625
Gene Name
zinc finger DHHC-type containing 7
Gene Aliases
DHHC7, SERZ-B, SERZ1, ZNF370
Chromosome Location
Chr. 16: 84974460 - 85011732 on Build GRCh38
UniGene ID
Hs.461610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145548.1 NP_001139020.1 8 1113
NM_017740.2 NP_060210.2 7 1002
XM_017023434.1 XP_016878923.1 5 538
XM_017023435.1 XP_016878924.1 5 398
XM_017023436.1 XP_016878925.1 5 513
GenBank mRNA AK000286.1 - 7 1002
AK001654.1 - 5 388
AK225768.1 - 7 954
BC017702.1 - 5 363 4394921
BC018772.1 - 8 1100 4869114

View Full Product Details