Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr. 1: 6634168 - 6701906 on Build GRCh38
UniGene ID
Hs.462640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018198.3 NP_060668.2 8 889
XM_017001745.1 XP_016857234.1 7 742
GenBank mRNA AF306694.1 - 8 788
AF306695.1 - 8 803
AK001599.1 - 8 798
AK294539.1 - 7 712
AK298378.1 - 8 775
AL109978.1 - 7 711
AL833841.1 - 8 763
AY336749.1 - 8 799
BC006086.1 - 8 785 3946414
BC008772.1 - 8 790
BC014145.2 - 8 797 4762069

View Full Product Details