Gene Transcripts

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
Chromosome Location
Chr. 6: 17759183 - 17987623 on Build GRCh38
UniGene ID
Hs.94499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105566.2 NP_001099036.1 7 672
NM_001105567.2 NP_001099037.1 7 672
NM_001105568.2 NP_001099038.1 7 672
NM_022113.5 NP_071396.4 7 672
XM_017011188.1 XP_016866677.1 7 667
XM_017011189.1 XP_016866678.1 7 667
XM_017011190.1 XP_016866679.1 7 667
XM_017011191.1 XP_016866680.1 5 699
XM_017011192.1 XP_016866681.1 7 667
XM_017011193.1 XP_016866682.1 4 575
GenBank mRNA AJ291578.1 - 7 512
AJ291579.1 - 7 512
AY014403.1 - 7 531
AY014404.1 - 7 531
BC127115.1 - 7 530 40125246
BC127116.1 - 7 530 40125251

View Full Product Details