Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022346.4 | NP_071741.2 | 10 | 1697 | |
NR_073124.1 | 9 | 1587 | |||
XM_011513876.2 | XP_011512178.1 | 9 | 1496 | ||
XM_017008543.1 | XP_016864032.1 | 10 | 1581 | ||
XM_017008544.1 | XP_016864033.1 | 9 | 1446 | ||
GenBank mRNA | AB013299.1 | - | 10 | 1571 | |
AF235023.1 | - | 10 | 1500 | ||
AF331796.1 | - | 10 | 1608 | ||
AK022512.1 | - | 10 | 1528 | ||
AK023147.1 | - | 9 | 1278 | ||
AK027511.1 | - | 1 | 34 | ||
BC068467.1 | - | 10 | 1514 | 4821684 | |
BC101476.1 | - | 10 | 1485 | 8068982 |