Gene Transcripts

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
Chromosome Location
Chr. 7: 100679507 - 100694282 on Build GRCh38
UniGene ID
Hs.414396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022574.4 NP_072096.2 13 2518
XM_005250532.1 XP_005250589.1 15 2299
XM_011516472.1 XP_011514774.1 15 2299
XM_011516477.1 XP_011514779.1 15 2299
XM_017012526.1 XP_016868015.1 15 2299
XM_017012527.1 XP_016868016.1 17 2197
XM_017012528.1 XP_016868017.1 16 1809
XM_017012529.1 XP_016868018.1 16 1816
XM_017012530.1 XP_016868019.1 16 2070
XM_017012531.1 XP_016868020.1 11 1322
GenBank mRNA AY176044.1 - 13 2719
BC129991.1 - 7 666

View Full Product Details