Gene Transcripts
- Gene Symbol
- MORC4
- Entrez Gene ID
- 79710
- Gene Name
- MORC family CW-type zinc finger 4
- Gene Aliases
- ZCW4, ZCWCC2, dJ75H8.2
- Chromosome Location
- Chr. X: 106940300 - 107000244 on Build GRCh38
- UniGene ID
- Hs.496544
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085354.2 |
NP_001078823.1
|
9, 10 |
1432 |
|
NM_024657.4 |
NP_078933.3
|
9, 10 |
1432 |
|
XM_005262190.3 |
XP_005262247.1
|
8, 9 |
1322 |
|
XM_006724691.2 |
XP_006724754.1
|
7, 8 |
1066 |
|
XM_011531027.2 |
XP_011529329.1
|
6, 7 |
720 |
|
XM_017029844.1 |
XP_016885333.1
|
8, 9 |
1322 |
|
GenBank mRNA |
AK056235.1 |
- |
5, 6 |
644 |
|
AK300305.1 |
- |
8, 9 |
1007 |
|
EF125209.1 |
- |
9, 10 |
1157 |
|
EF125210.1 |
- |
8, 9 |
1216 |
|
View Full Product Details