Gene Transcripts

Gene Symbol
ASRGL1
Entrez Gene ID
80150
Gene Name
asparaginase like 1
Gene Aliases
ALP, ALP1, CRASH
Chromosome Location
Chr. 11: 62337302 - 62401399 on Build GRCh38
UniGene ID
Hs.535326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083926.1 NP_001077395.1 7 1160
NM_025080.3 NP_079356.3 7 1083
XM_005274305.3 XP_005274362.1 6 800
XM_005274306.2 XP_005274363.1 6 862
GenBank mRNA AF411076.1 - 7 892
AK090908.1 - 7 932
AK091894.1 - 7 1014
AK225802.1 - 7 851
AK313069.1 - 7 943
BC006267.2 - 4 473
BC021295.2 - 7 881 3937243
BC064963.1 - 6 773 6141919
BC093070.1 - 7 985 30370741
BX640832.1 - 1 3180

View Full Product Details