Gene Transcripts

Gene Symbol
GPR63
Entrez Gene ID
81491
Gene Name
G protein-coupled receptor 63
Gene Aliases
PSP24(beta), PSP24B
Chromosome Location
Chr. 6: 96794122 - 96837477 on Build GRCh38
UniGene ID
Hs.221364
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143957.2 NP_001137429.1 3 1022
NM_030784.3 NP_110411.1 2 904
XM_006715570.3 XP_006715633.1 2 974
XM_011536153.2 XP_011534455.1 3 965
XM_011536154.2 XP_011534456.1 3 838
XM_011536155.2 XP_011534457.1 3 1017
XM_011536157.2 XP_011534459.1 2 740
XM_011536159.2 XP_011534461.1 2 909
XM_017011334.1 XP_016866823.1 2 1432
GenBank mRNA AB030566.1 - 2 751
AB209347.1 - 2 890
AK289839.1 - 2 904
AK313535.1 - 2 873
BC067465.1 - 1 579 6971763
BC067466.1 - 1 580 6971764
BC067467.1 - 1 580 6971765
BC067468.1 - 1 580 6971766
BC067469.1 - 1 580 6971767
FJ348260.1 - 1 544

View Full Product Details