Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000766.4 | NP_000757.2 | 9 | 1391 | |
GenBank mRNA | AF209774.1 | - | 9 | 1370 | |
AY513604.1 | - | 9 | 1370 | ||
AY513605.1 | - | 9 | 1370 | ||
AY513607.1 | - | 9 | 1370 | ||
AY513608.1 | - | 9 | 1370 | ||
AY513609.1 | - | 9 | 1370 |