Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr. 1: 1292384 - 1307925 on Build GRCh38
UniGene ID
Hs.535257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030649.2 NP_085152.2 5, 6 405
XM_005244715.2 XP_005244772.1 5, 6 441
XM_011540606.2 XP_011538908.1 5, 6 454
XM_011540607.1 XP_011538909.1 5, 6 2878
XM_011540608.2 XP_011538910.1 5, 6 455
XM_011540609.2 XP_011538911.1 5, 6 455
GenBank mRNA AB051503.1 - 4, 5 2070
AF411981.1 - 3, 4 205
AK093954.1 - 5, 6 452
BC051194.1 - 5, 6 405 4819254

View Full Product Details