Gene Transcripts

Gene Symbol
SPPL3
Entrez Gene ID
121665
Gene Name
signal peptide peptidase like 3
Gene Aliases
IMP2, MDHV1887, PRO4332, PSH1, PSL4
Chromosome Location
Chr. 12: 120762510 - 120904352 on Build GRCh38
UniGene ID
Hs.507087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139015.4 NP_620584.2 9 1309
XM_011537925.2 XP_011536227.1 9 839
GenBank mRNA AB252457.1 - 9 901
AJ345030.1 - 9 877
AJ420898.1 - 9 848
AK074916.1 - 8 792
AK223590.1 - 9 1305
AK291161.1 - 9 1131
AY169313.1 - 9 1083
AY358767.1 - 2 155
BC009551.1 - 2 177
BC025781.1 - 6 584
BC073910.1 - 9 1090 4538522
BC101625.1 - 9 904 8069131
BC101627.1 - 9 904 8069133

View Full Product Details