Gene Transcripts

Gene Symbol
TMCO2
Entrez Gene ID
127391
Gene Name
transmembrane and coiled-coil domains 2
Gene Aliases
dJ39G22.2
Chromosome Location
Chr. 1: 40247901 - 40251693 on Build GRCh38
UniGene ID
Hs.406265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008740.3 NP_001008740.1 2 435
GenBank mRNA BC108666.1 - 2 368
BU567634.1 - 2 438
BU569147.1 - 2 404
DB518323.1 - 2 136

View Full Product Details