Gene Transcripts
- Gene Symbol
- HGSNAT
- Entrez Gene ID
- 138050
- Gene Name
- heparan-alpha-glucosaminide N-acetyltransferase
- Gene Aliases
- HGNAT, MPS3C, RP73, TMEM76
- Chromosome Location
- Chr. 8: 43140449 - 43202827 on Build GRCh38
- UniGene ID
- Hs.600384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152419.2 |
NP_689632.2
|
3, 4 |
420 |
|
XM_005273409.1 |
XP_005273466.1
|
3, 4 |
420 |
|
XM_005273410.1 |
XP_005273467.1
|
3, 4 |
420 |
|
XM_005273411.1 |
XP_005273468.1
|
3, 4 |
420 |
|
XM_005273412.3 |
XP_005273469.1
|
3, 4 |
420 |
|
XM_017013088.1 |
XP_016868577.1
|
3, 4 |
420 |
|
XM_017013089.1 |
XP_016868578.1
|
3, 4 |
324 |
|
GenBank mRNA |
AK304441.1 |
- |
3, 4 |
372 |
|
BC042037.1 |
- |
3, 4 |
365 |
|
BE327420.1 |
- |
2, 3 |
268 |
|
DA395558.1 |
- |
3, 4 |
374 |
|
DB128369.1 |
- |
3, 4 |
420 |
|
View Full Product Details