Gene Transcripts

Gene Symbol
EGR3
Entrez Gene ID
1960
Gene Name
early growth response 3
Gene Aliases
EGR-3, PILOT
Chromosome Location
Chr. 8: 22687661 - 22693302 on Build GRCh38
UniGene ID
Hs.534313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004430.2 NP_004421.2 1 485
XM_005273426.3 XP_005273483.1 1 56
GenBank mRNA AK292464.1 - 1 381
AK313604.1 - 1 381
BC104765.1 - 1 226 8143768
BC112279.1 - 1 224 8327747
S40832.1 - 1 484
X63741.1 - 1 480

View Full Product Details