Gene Transcripts
- Gene Symbol
- SMIM14
- Entrez Gene ID
- 201895
- Gene Name
- small integral membrane protein 14
- Gene Aliases
- C4orf34
- Chromosome Location
- Chr. 4: 39547297 - 39638998 on Build GRCh38
- UniGene ID
- Hs.205952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317896.1 |
NP_001304825.1
|
3, 4 |
513 |
|
NM_001317897.1 |
NP_001304826.1
|
2, 3 |
222 |
|
NM_174921.2 |
NP_777581.1
|
2, 3 |
368 |
|
XM_011513640.2 |
XP_011511942.1
|
2, 3 |
501 |
|
XM_017007862.1 |
XP_016863351.1
|
3, 4 |
417 |
|
XM_017007863.1 |
XP_016863352.1
|
2, 3 |
258 |
|
GenBank mRNA |
AK023351.1 |
- |
2, 3 |
231 |
|
BC008502.1 |
- |
2, 3 |
231 |
4295121 |
BQ049060.1 |
- |
2, 3 |
213 |
|
HY004916.1 |
- |
3, 4 |
414 |
|
HY155982.1 |
- |
2, 3 |
369 |
|
View Full Product Details