Gene Transcripts

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
Chromosome Location
Chr. 11: 101890674 - 101916522 on Build GRCh38
UniGene ID
Hs.318370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178127.4 NP_835228.2 5 955
GenBank mRNA AK291523.1 - 4 419
AY169281.1 - 4 413
AY358827.1 - 4 403
BC049170.1 - 5 958 5285447
BC053957.1 - 5 958

View Full Product Details