Gene Transcripts
- Gene Symbol
- TMEM86B
- Entrez Gene ID
- 255043
- Gene Name
- transmembrane protein 86B
- Gene Aliases
- Chromosome Location
- Chr. 19: 55226634 - 55229264 on Build GRCh38
- UniGene ID
- Hs.135215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC023000.1 |
- |
3 |
1610 |
|
BC033193.1 |
- |
2 |
1372 |
|
View Full Product Details