Gene Transcripts

Gene Symbol
C17orf51
Entrez Gene ID
339263
Gene Name
chromosome 17 open reading frame 51
Gene Aliases
Chromosome Location
Chr. 17: 21524606 - 21551678 on Build GRCh38
UniGene ID
Hs.514016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113434.4 NP_001106905.1 1 668
XM_005256621.3 XP_005256678.1 1 661
XM_006721514.2 XP_006721577.1 1 661
XM_011523822.2 XP_011522124.1 1 661
GenBank mRNA AK056436.1 - 1 634
BC010612.1 - 1 666
BC019938.1 - 1 656
BC052777.1 - 1 668
BC053952.1 - 1 668
BC132738.1 - 1 642 40146760
BC136630.1 - 1 642 9020620

View Full Product Details