Gene Transcripts

Gene Symbol
SMTNL2
Entrez Gene ID
342527
Gene Name
smoothelin like 2
Gene Aliases
Chromosome Location
Chr. 17: 4583981 - 4608319 on Build GRCh38
UniGene ID
Hs.441709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114974.1 NP_001108446.1 3, 4 796
NM_198501.2 NP_940903.2 3, 4 448
XM_006721515.2 XP_006721578.1 3, 4 964
GenBank mRNA AK124452.1 - 3, 4 430
BC105050.1 - 3, 4 349 8144053
BC110859.1 - 3, 4 796 5190568
BC112177.1 - 3, 4 349 8327645
DA822419.1 - 3, 4 448

View Full Product Details