Gene Transcripts

Gene Symbol
OR2T5
Entrez Gene ID
401993
Gene Name
olfactory receptor family 2 subfamily T member 5
Gene Aliases
OR1-62
Chromosome Location
Chr. 1: 248488589 - 248489536 on Build GRCh38
UniGene ID
Hs.553808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004697.1 NP_001004697.1 1 878
GenBank mRNA BC140747.1 - 1 883
BC140915.1 - 1 903 9056727

Gene Symbol
OR2T29
Entrez Gene ID
343563
Gene Name
olfactory receptor family 2 subfamily T member 29
Gene Aliases
Chromosome Location
Chr. 1: 248558544 - 248559491 on Build GRCh38
UniGene ID
Hs.553707
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004694.2 NP_001004694.2 1 878

Gene Symbol
OR2T4
Entrez Gene ID
127074
Gene Name
olfactory receptor family 2 subfamily T member 4
Gene Aliases
OR1-60, OR2T4Q
Chromosome Location
Chr. 1: 248361581 - 248362627 on Build GRCh38
UniGene ID
Hs.553586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004696.1 NP_001004696.1 1 962

View Full Product Details